Learn More
1-Octyl isocyanate, 97%, Thermo Scientific Chemicals
$103.11 - $103.11
Chemical Identifiers
| CAS | 3158-26-7 |
|---|---|
| Molecular Formula | C9H17NO |
| Molecular Weight (g/mol) | 155.241 |
| MDL Number | MFCD00013887 |
| InChI Key | DYQFCTCUULUMTQ-UHFFFAOYSA-N |
| Synonym | octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate |
| PubChem CID | 76616 |
| IUPAC Name | 1-isocyanatooctane |
| SMILES | CCCCCCCCN=C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1874606
|
Thermo Scientific Chemicals
L1874606 |
5 g |
Each for $103.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3158-26-7 | |
| 155.241 | |
| DYQFCTCUULUMTQ-UHFFFAOYSA-N | |
| 76616 | |
| CCCCCCCCN=C=O |
| C9H17NO | |
| MFCD00013887 | |
| octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate | |
| 1-isocyanatooctane |
Specifications
| 3158-26-7 | |
| 200°C to 204°C | |
| Pungent | |
| 1.432 | |
| 5 g | |
| Moisture sensitive | |
| DYQFCTCUULUMTQ-UHFFFAOYSA-N | |
| 1-isocyanatooctane | |
| 76616 | |
| 97% |
| 0.88 | |
| 71°C (159°F) | |
| C9H17NO | |
| MFCD00013887 | |
| UN2206 | |
| octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate | |
| CCCCCCCCN=C=O | |
| 155.241 | |
| 155.24 | |
| 1-Octyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 221-598-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only