missing translation for 'onlineSavingsMsg'
Learn More

1-Octene 97.0+%, TCI America™

Catalog No. p-7134149
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
O004125ML 25 mL
O0041500ML 500 mL
2 options

Catalog No. O004125ML

Supplier: TCI America O004125ML

Only null left

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

Melting Point -101°C
Color Colorless
Boiling Point 123°C
Quantity 25 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octene
EINECSNumber (2)-0024
RTECSNumber RH2207000
TSCA Yes
Recommended Storage Refrigerator