missing translation for 'onlineSavingsMsg'
Learn More

1-Octene 97.0+%, TCI America™

Catalog No. O0041500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0041500ML 500 mL
O004125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0041500ML Supplier TCI America Supplier No. O0041500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

Melting Point -101°C
Color Colorless
Boiling Point 123°C
Quantity 500 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octene
EINECSNumber (2)-0024
RTECSNumber RH2207000
TSCA Yes
Recommended Storage Refrigerator