missing translation for 'onlineSavingsMsg'
Learn More

1-Octene 99.5+%, TCI America™

Catalog No. S03385ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03385ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03385ML Supplier TCI America Supplier No. S03385ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

Color Colorless
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octene [Standard Material for GC]
EINECSNumber (2)-0024
RTECSNumber RH2207000
TSCA Yes