Learn More
1-Octene, 99+%
CAS: 111-66-0 | C8H16 | 112.21 g/mol
Supplier: Thermo Scientific Chemicals 301250025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Octene | |
| 111-66-0 | |
| 100.0 | |
| 0.7150g/mL | |
| 13°C | |
| 99% min. (GC) | |
| C8H16 | |
| CH3(CH2)5CH=CH2 | |
| 2.5 L | |
| 0.715 | |
| 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene | |
| KWKAKUADMBZCLK-UHFFFAOYSA-N | |
| oct-1-ene | |
| 8125 | |
| CHEBI:46708 | |
| 99+% |
| 99+% | |
| 99.0 | |
| -102.0°C | |
| 121.0°C to 123.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4080 to 1.4100 | |
| MFCD00009548 | |
| 01, 221 | |
| 15, 1766 | |
| Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents | |
| CCCCCCC=C | |
| 112.21 | |
| 0.38 mm2/s (40°C) | |
| 112.21 | |
| Liquid |
Chemical Identifiers
| 111-66-0 | |
| 112.21 | |
| KWKAKUADMBZCLK-UHFFFAOYSA-N | |
| 8125 | |
| oct-1-ene |
| C8H16 | |
| MFCD00009548 | |
| 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene | |
| CHEBI:46708 | |
| CCCCCCC=C |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environm
GHS Signal Word: Danger
EINECSNumber : 203-893-7
TSCA : TSCA
RUO – Research Use Only