missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, 99+%

Catalog No. AC301255000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301255000 500 mL Glass bottle
AC301251000 100 mL Glass bottle
AC301250010 1 L Glass bottle
AC301250025 2.5 L Glass bottle
AC301250100 10 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC301255000 Supplier Thermo Scientific Chemicals Supplier No. 301255000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-66-0 | C8H16 | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -102.0°C
Density 0.7150g/mL
Boiling Point 121.0°C to 123.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH=CH2
Refractive Index 1.4080 to 1.4100
Quantity 500 mL
Beilstein 01, 221
Specific Gravity 0.715
Merck Index 15, 1766
Solubility Information Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents
Viscosity 0.38 mm2/s (40°C)
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Octene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environm
EINECSNumber 203-893-7
TSCA TSCA

RUO – Research Use Only