missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-one 95.0+%, TCI America™

Catalog No. p-7134306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03821G 1 g
O03825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03821G Supplier TCI America Supplier No. O03821G
Only null left

Chemical Identifiers

CAS 4312-99-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00036558
InChI Key KLTVSWGXIAYTHO-UHFFFAOYSA-N
Synonym 1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
PubChem CID 61346
IUPAC Name oct-1-en-3-one
SMILES CCCCCC(=O)C=C

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 1224
Formula Weight 126.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octen-3-one
TSCA Yes
Recommended Storage Refrigerator