missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-one, 97%, stab. with 0.1% BHA

Catalog No. AAL0718903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0718903 1 g
AAL0718906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0718903 Supplier Thermo Scientific Chemicals Supplier No. L0718903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4312-99-6 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4312-99-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00036558
InChI Key KLTVSWGXIAYTHO-UHFFFAOYSA-N
Synonym 1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
PubChem CID 61346
IUPAC Name oct-1-en-3-one
SMILES CCCCCC(=O)C=C

Specifications

Density 0.848
Boiling Point 56°C to 60°C (16 mmHg)
Flash Point 35°C (95°F)
Odor Characteristic
Refractive Index 1.436
Quantity 1 g
UN Number UN1224
Beilstein 1700905
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1-Octen-3-one, stabilized with 0.1% BHA
Show More Show Less
Hazard Category H226-H302-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 224-327-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only