missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%

Catalog No. p-7081898
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1207909 10 g
AAA1207918 50 g
AAA1207930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1207909 Supplier Thermo Scientific Chemicals Supplier No. A1207909

CAS: 3391-86-4 | C8H16O | 128.215 g/mol

Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00004589
InChI Key VSMOENVRRABVKN-UHFFFAOYSA-N
Synonym 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
PubChem CID 18827
ChEBI CHEBI:34118
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(C=C)O

Specifications

Melting Point -49°C
Density 0.833
Boiling Point 173°C to 175°C
Flash Point 61°C (141°F)
Odor Characteristic
Refractive Index 1.437
Quantity 10 g
Beilstein 1744110
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Octen-3-ol
Show More Show Less
Hazard Category H227-H302-H315-H319-H332
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
EINECSNumber 222-226-0
RTECSNumber RH3300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only