missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol 99.0+%, TCI America™

Catalog No. O003625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003625ML 25 mL
O0036500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003625ML Supplier TCI America Supplier No. O003625ML

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 194°C
Quantity 25 mL
Formula Weight 130.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octanol
EINECSNumber (2)-0217
RTECSNumber RH6550000
TSCA Yes