missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, Reagents
Spécifications
Spécifications
| Boiling Range | 196°C |
| Melting Point | -18°C |
| Color | Colorless |
| Boiling Point | 196°C |
| Melting Range | -18°C |
| Packaging | Glass Bottle |
| Quantity | 100 g |
| Specific Gravity | 0.827 |
| Grade | Reagent |
| Physical Form | Liquid |
| Afficher plus de résultats |