missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, ReagentPlus™, 99%, Honeywell™
Reagent Grade, 99%
$319.32
Chemical Identifiers
| CAS | 111-87-5 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00002988 |
| InChI Key | KBPLFHHGFOOTCA-UHFFFAOYSA-N |
| Synonym | 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol |
| PubChem CID | 957 |
| ChEBI | CHEBI:16188 |
| IUPAC Name | octan-1-ol |
| SMILES | CCCCCCCCO |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| C8H18O | |
| MFCD00002988 | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| CHEBI:16188 | |
| CCCCCCCCO |
Specifications
| 111-87-5 | |
| Colorless | |
| 196°C | |
| C8H18O | |
| CH3(CH2)7OH | |
| 1697461 | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| octan-1-ol | |
| 957 | |
| CHEBI:16188 | |
| 99% | |
| Liquid |
| -15°C | |
| 0.827g/mL at 25°C | |
| 80°C (176°F) | |
| n20/D 1.429(lit.) | |
| MFCD00002988 | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| CCCCCCCCO | |
| 130.23 | |
| 4.5 (vs air) | |
| 130.23g/mol | |
| ReagentPlus | |
| 1-Octanol |
Safety and Handling
P273-P280-P305 + P351 + P338-P337 + P313
H319-H412
EINECSNumber : 203-917-6