missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol 99.5+%, TCI America™

Catalog No. O0212500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0212500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0212500ML Supplier TCI America Supplier No. O0212500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -16°C
Color Colorless
Boiling Point 194°C
Quantity 500 mL
Formula Weight 130.23
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octanol [for Determining Partition Coefficients]
EINECSNumber (2)-0217
RTECSNumber RH6550000
TSCA Yes