missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ACS reagent, ≥99%, Solstice

Catalog No. 60046951
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046951 100 mL Glass Bottle
60046952 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046951 Supplier Solstice Supplier No. 472328100ML

ACS Reagent,≥99%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Density 0.827g/mL at 25°C
Boiling Point 194°C
Flash Point 80°C (176°F)
Odor Characteristic
Packaging Glass Bottle
Refractive Index n20/D 1.429
Linear Formula CH3(CH2)7OH
Quantity 100 mL
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity ≥99%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P337 + P313
RTECSNumber 203-917-6