Learn More
1-Octanol, ACS reagent
CAS: 111-87-5 | C8H18O | 130.23 g/mol
$172.96 - $172.96
Chemical Identifiers
| CAS | 111-87-5 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00002988 |
| InChI Key | KBPLFHHGFOOTCA-UHFFFAOYSA-N |
| Synonym | 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol |
| PubChem CID | 957 |
| ChEBI | CHEBI:16188 |
| IUPAC Name | octan-1-ol |
| SMILES | CCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC220180010
|
Thermo Scientific Chemicals
220180010 |
1 L | Glass bottle |
Each for $172.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111-87-5 | |
| 130.23 | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| 957 | |
| octan-1-ol |
| C8H18O | |
| MFCD00002988 | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| CHEBI:16188 | |
| CCCCCCCCO |
Specifications
| 111-87-5 | |
| 100.0 | |
| 0.8240g/mL | |
| 99+% | |
| C8H18O | |
| CH3(CH2)7OH | |
| 1 L | |
| 0.004 % max. | |
| 15, 6840 | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| octan-1-ol | |
| 957 | |
| 130.23 | |
| ACS Reagent | |
| 1-Octanol, 99+% |
| 99.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| 1.4280 to 1.4310 (20°C, 589nm) | |
| MFCD00002988 | |
| 01, 418 | |
| 0.822 to 0.826 (25°/25°C) | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| CCCCCCCCO | |
| 130.23 | |
| CHEBI:16188 | |
| ≥99.0% (GC) | |
| Fine Crystalline Powder |
Safety and Handling
GHS P Statement Causes serious eye irritation. Harmful to aquatic life with long lasting effects.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Warning
EINECSNumber : 203-917-6
RTECSNumber : RH6550000
TSCA : TSCA
RUO – Research Use Only