missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ACS reagent

Catalog No. AC220180010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220180010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC220180010 Supplier Thermo Scientific Chemicals Supplier No. 220180010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-87-5 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Density 0.8240g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4280 to 1.4310 (20°C, 589nm)
Linear Formula CH3(CH2)7OH
Quantity 1 L
Beilstein 01, 418
Residue after Evaporation 0.004 % max.
Merck Index 15, 6840
Specific Gravity 0.822 to 0.826 (25°/25°C)
Formula Weight 130.23
Percent Purity ≥99.0% (GC)
Grade ACS Reagent
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Octanol, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation. Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
EINECSNumber 203-917-6
RTECSNumber RH6550000
TSCA TSCA

RUO – Research Use Only