missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, 99%

CAS: 111-87-5 | C8H18O | 130.23 g/mol

Supplier:  Thermo Scientific Chemicals A159770F

Quantity 2500 mL

 View more versions of this product

Catalog No. AAA159770F


Only null left

Description

Description

1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

111-87-5
130.23
KBPLFHHGFOOTCA-UHFFFAOYSA-N
957
octan-1-ol
C8H18O
MFCD00002988
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
CHEBI:16188
CCCCCCCCO

Specifications

1-Octanol
-16°C
193°C to 195°C
Sweet
C8H18O
CH3(CH2)7OH
MFCD00002988
14,6751
Miscible with ethanol,ether and propylene glycol. Immiscible with water.
CCCCCCCCO
130.23
CHEBI:16188
99%
111-87-5
0.825
81°C (177°F)
99%
1.429
2500 mL
1697461
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
KBPLFHHGFOOTCA-UHFFFAOYSA-N
octan-1-ol
957
130.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

EINECSNumber : 203-917-6

RTECSNumber : RH6550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions