Learn More
1-Octanol, 99%
CAS: 111-87-5 | C8H18O | 130.23 g/mol
Supplier: Thermo Scientific Chemicals A159770D
| Quantity | 10,000 mL |
|---|
Description
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.
Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| C8H18O | |
| MFCD00002988 | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| CHEBI:16188 | |
| CCCCCCCCO |
Specifications
| 1-Octanol | |
| -16°C | |
| 193°C to 195°C | |
| Sweet | |
| C8H18O | |
| CH3(CH2)7OH | |
| MFCD00002988 | |
| 14,6751 | |
| Miscible with ethanol,ether and propylene glycol. Immiscible with water. | |
| CCCCCCCCO | |
| 130.23 | |
| CHEBI:16188 | |
| 99% |
| 111-87-5 | |
| 0.825 | |
| 81°C (177°F) | |
| 99% | |
| 1.429 | |
| 10,000 mL | |
| 1697461 | |
| 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol | |
| KBPLFHHGFOOTCA-UHFFFAOYSA-N | |
| octan-1-ol | |
| 957 | |
| 130.23 |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
H227-H319
EINECSNumber : 203-917-6
RTECSNumber : RH6550000
TSCA : Yes
Recommended Storage : Ambient temperatures