missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonyl Chloride 95.0+%, TCI America™
$72.70 - $784.76
Chemical Identifiers
| CAS | 7795-95-1 |
|---|---|
| Molecular Formula | C8H17ClO2S |
| Molecular Weight (g/mol) | 212.732 |
| MDL Number | MFCD00014729 |
| InChI Key | WIVNTNLDTMNDNO-UHFFFAOYSA-N |
| Synonym | 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx |
| PubChem CID | 82261 |
| IUPAC Name | octane-1-sulfonyl chloride |
| SMILES | CCCCCCCCS(=O)(=O)Cl |
Chemical Identifiers
| 7795-95-1 | |
| 212.732 | |
| WIVNTNLDTMNDNO-UHFFFAOYSA-N | |
| 82261 | |
| CCCCCCCCS(=O)(=O)Cl |
| C8H17ClO2S | |
| MFCD00014729 | |
| 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx | |
| octane-1-sulfonyl chloride |
Specifications
| 7795-95-1 | |
| C8H17ClO2S | |
| 25 g | |
| 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx | |
| CCCCCCCCS(=O)(=O)Cl | |
| 212.732 | |
| 212.73 | |
| Liquid |
| 108°C | |
| MFCD00014729 | |
| 3265 | |
| WIVNTNLDTMNDNO-UHFFFAOYSA-N | |
| octane-1-sulfonyl chloride | |
| 82261 | |
| ≥95.0% (GC,T) | |
| 1-Octanesulfonyl Chloride |
Safety and Handling
EINECSNumber : (2)-1641
TSCA : Yes