missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonyl Chloride 95.0+%, TCI America™

Catalog No. O015025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O015025G 25 g
O0150500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O015025G Supplier TCI America Supplier No. O015025G

Chemical Identifiers

CAS 7795-95-1
Molecular Formula C8H17ClO2S
Molecular Weight (g/mol) 212.732
MDL Number MFCD00014729
InChI Key WIVNTNLDTMNDNO-UHFFFAOYSA-N
Synonym 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx
PubChem CID 82261
IUPAC Name octane-1-sulfonyl chloride
SMILES CCCCCCCCS(=O)(=O)Cl

Specifications

Boiling Point 108°C
Quantity 25 g
UN Number 3265
Formula Weight 212.73
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Octanesulfonyl Chloride
EINECSNumber (2)-1641
TSCA Yes