missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC, 99%, Spectrum™ Chemical
O1001, 207596-29-0, C8H17NaO3S·H2O
Supplier: Spectrum Chemical Mfg Cor O1001100GM
Description
Spectrum™ Chemical 1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC is an ion pairing reagent which will enhance the retention of polar, ionizable compounds such as peptides and proteins. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Specifications
| 207596-29-0 | |
| Amber Glass Bottle | |
| MFCD00149551 | |
| MBURIAHQXJQKRE-UHFFFAOYSA-M | |
| sodium hydrate octane-1-sulfonate | |
| 99% |
| 100% | |
| C8H19NaO4S | |
| 100 g | |
| O.[Na+].CCCCCCCCS([O-])(=O)=O | |
| 234.29 | |
| Reagent |
Chemical Identifiers
| 207596-29-0 | |
| 234.29 | |
| MBURIAHQXJQKRE-UHFFFAOYSA-M | |
| O.[Na+].CCCCCCCCS([O-])(=O)=O |
| C8H19NaO4S | |
| MFCD00149551 | |
| sodium hydrate octane-1-sulfonate |