missing translation for 'onlineSavingsMsg'
Learn More

1 Octane Sulfonic Acid Sodium Salt, HPLC Ion Pair Agent, Fisher Chemical™

Catalog No. p-8820003
Change view
Click to view available options
Quantity:
100 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002825 25 g
O0028100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O002825 Supplier Thermo Fisher Scientific Supplier No. O002825

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve HPLC separation of basic analytes by ion pairing.

  • High purity
  • Application tested
  • Dissolves easily

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
IUPAC Name sodium octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Absorbance ≤ 1.00 AU at 210 nm, ≤ 0.1 AU at 220 nm, ≤ 0.04 AU at 254 nm, ≤ 0.04 AU at 280nm
Loss on Drying 1.5% Max.
Infrared Spectrum Conforms
Quantity 25 g
Formula Weight 216.7
Grade HPLC
Physical Form Powder
Recommended Storage Ambient