missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecyne 95.0+%, TCI America™

Catalog No. p-7134200
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01281ML 1 mL
O01285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01281ML Supplier TCI America Supplier No. O01281ML
Only null left

Chemical Identifiers

CAS 629-89-0
Molecular Formula C18H34
Molecular Weight (g/mol) 250.47
MDL Number MFCD00015088
InChI Key IYDNQWWOZQLMRH-UHFFFAOYSA-N
Synonym 1-octadecyne, octadecyne, octadec-1-in, cetylacetylene, hexadecylacetylene, acmc-1b2rb, qspl 036
PubChem CID 69425
IUPAC Name octadec-1-yne
SMILES CCCCCCCCCCCCCCCCC#C

Specifications

Color White-Yellow
Quantity 1 mL
Formula Weight 250.47
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Octadecyne