missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecyne, 96%

Catalog No. AAH3328809
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3328809 10 g
AAH3328803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3328809 Supplier Thermo Scientific Chemicals Supplier No. H3328809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-89-0 | C18H34 | 250.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-89-0
Molecular Formula C18H34
Molecular Weight (g/mol) 250.47
MDL Number MFCD00015088
InChI Key IYDNQWWOZQLMRH-UHFFFAOYSA-N
Synonym 1-octadecyne, octadecyne, octadec-1-in, cetylacetylene, hexadecylacetylene, acmc-1b2rb, qspl 036
PubChem CID 69425
IUPAC Name octadec-1-yne
SMILES CCCCCCCCCCCCCCCCC#C

Specifications

Melting Point 21°C to 24°C
Density 0.803
Boiling Point 313°C
Quantity 10 g
Formula Weight 250.47
Percent Purity ≥97%
Chemical Name or Material 1-Octadecyne
EINECSNumber 211-114-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only