missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylpyrrole 95.0+%, TCI America™

Catalog No. O02195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02195G 5 g
O02191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02195G Supplier TCI America Supplier No. O02195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89601-24-1
Molecular Formula C22H41N
Molecular Weight (g/mol) 319.577
MDL Number MFCD00144990
InChI Key BMSMIJNAHWEOBQ-UHFFFAOYSA-N
PubChem CID 19013669
IUPAC Name 1-octadecylpyrrole
SMILES CCCCCCCCCCCCCCCCCCN1C=CC=C1

Specifications

Melting Point 39°C
Color White
Boiling Point 182°C
Quantity 5 g
Formula Weight 319.58
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Octadecylpyrrole
TSCA No
Recommended Storage Refrigerator