missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene 90.0+%, TCI America™

Catalog No. O000825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O000825ML 25 mL
O0008500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O000825ML Supplier TCI America Supplier No. O000825ML
Only null left

Chemical Identifiers

CAS 112-88-9
Molecular Formula C18H36
Molecular Weight (g/mol) 252.486
MDL Number MFCD00009003
InChI Key CCCMONHAUSKTEQ-UHFFFAOYSA-N
Synonym 1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
PubChem CID 8217
ChEBI CHEBI:30824
IUPAC Name octadec-1-ene
SMILES CCCCCCCCCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 179°C
Quantity 25 mL
Formula Weight 252.49
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Octadecene
EINECSNumber (2)-0027
TSCA Yes