missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene, 90%, tech.

Catalog No. p-4509848
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129310010 1 L Glass bottle
AC129310025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129310010 Supplier Thermo Scientific Chemicals Supplier No. 129310010
Only null left

CAS: 112-88-9 | C18H36 | 252.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-88-9
Molecular Formula C18H36
Molecular Weight (g/mol) 252.48
InChI Key CCCMONHAUSKTEQ-UHFFFAOYSA-N
Synonym 1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
PubChem CID 8217
ChEBI CHEBI:30824
IUPAC Name octadec-1-ene
SMILES CCCCCCCCCCCCCCCCC=C

Specifications

Melting Point 17.5°C
Color Colorless to Amber
Density 0.7890g/mL
Boiling Point 315°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Refractive Index 1.4435 to 1.4455
Linear Formula CH3(CH2)15CH=CH2
Quantity 1 L
Beilstein 01,226
Specific Gravity 0.789
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents
Viscosity 3.6 mm2/s (40°C)
Formula Weight 252.48
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Octadecene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 204-012-9

RUO – Research Use Only