missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol 98.0+%, TCI America™

Catalog No. p-7134120
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O000625G 25 g
O0006500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O000625G Supplier TCI America Supplier No. O000625G
Only null left

Chemical Identifiers

CAS 112-92-5
Molecular Formula C18H38O
Molecular Weight (g/mol) 270.501
MDL Number MFCD00002823
InChI Key GLDOVTGHNKAZLK-UHFFFAOYSA-N
Synonym stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
PubChem CID 8221
ChEBI CHEBI:32154
IUPAC Name octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 59°C
Color White
Boiling Point 210°C
Quantity 25 g
Formula Weight 270.50
Percent Purity ≥98.0% (GC)
Physical Form Pellets
Chemical Name or Material 1-Octadecanol
EINECSNumber (2)-0217
RTECSNumber RG2010000
TSCA Yes