missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanol, 97%

CAS: 112-92-5 | C18H38O | 270.501 g/mol

Supplier:  Thermo Scientific Chemicals A120200E

 View more versions of this product

Catalog No. AAA120200E


Only null left
Explore available promotions

Description

Description

1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Specifications

Specifications

1-Octadecanol
56°C to 60°C
174°C (345°F)
C18H38O
2500 g
14,8805
Insoluble in water. Soluble in alcohol,acetone,ether,benzene,chloroform.
CCCCCCCCCCCCCCCCCCO
270.501
CHEBI:32154
97%
112-92-5
210°C (15 mmHg)
97%
MFCD00002823
1362907
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
GLDOVTGHNKAZLK-UHFFFAOYSA-N
octadecan-1-ol
8221
270.5

Chemical Identifiers

112-92-5
270.501
GLDOVTGHNKAZLK-UHFFFAOYSA-N
8221
octadecan-1-ol
C18H38O
MFCD00002823
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
CHEBI:32154
CCCCCCCCCCCCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-017-6

RTECSNumber : RG2010000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only