missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-Octadecanol

Catalog No. 258768100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
258768100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 258768100G Supplier Sigma Aldrich Supplier No. 258768100G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octadecanol

Specifications

CAS 112-92-5
Melting Point 56°C to 59°C (lit.)
Density 0.812 g/mL (at 25°C (literature))
Boiling Point 170°C to 171°C (2 mmHg)
Molecular Formula C18H38O
Linear Formula CH3(CH2)17 OH
MDL Number MFCD00002823
Quantity 100 g
Synonym Octadecyl alcohol; Stearyl alcohol
Molecular Weight (g/mol) 270.49
Percent Purity 99%
Show More Show Less