missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Octadecanethiol 97.0+%, TCI America™
$44.79 - $273.73
Chemical Identifiers
CAS | 2885-00-9 |
---|---|
Molecular Formula | C18H38S |
Molecular Weight (g/mol) | 286.56 |
MDL Number | MFCD00004886 |
InChI Key | QJAOYSPHSNGHNC-UHFFFAOYSA-N |
Synonym | octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan |
PubChem CID | 17905 |
IUPAC Name | octadecane-1-thiol |
SMILES | CCCCCCCCCCCCCCCCCCS |
Chemical Identifiers
2885-00-9 | |
286.56 | |
QJAOYSPHSNGHNC-UHFFFAOYSA-N | |
17905 | |
CCCCCCCCCCCCCCCCCCS |
C18H38S | |
MFCD00004886 | |
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan | |
octadecane-1-thiol |
Specifications
2885-00-9 | |
C18H38S | |
25 g | |
QJAOYSPHSNGHNC-UHFFFAOYSA-N | |
octadecane-1-thiol | |
17905 | |
≥97.0% (GC,T) | |
1-Octadecanethiol |
White | |
MFCD00004886 | |
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan | |
CCCCCCCCCCCCCCCCCCS | |
286.56 | |
286.56 | |
Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes