missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanethiol, 96%

CAS: 2885-00-9 | C18H38S | 286.56 g/mol

Supplier:  Thermo Scientific Chemicals L0412314

 View more versions of this product

Catalog No. AAL0412314


Only null left
Explore available promotions

Description

Description

1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

1-Octadecanethiol
30°C to 33°C
180°C (5 mmHg)
Stench
MFCD00004886
1811934
Insoluble in water.
CCCCCCCCCCCCCCCCCCS
286.56
286.57
2885-00-9
0.847
185°C (365°F)
C18H38S
25 g
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
QJAOYSPHSNGHNC-UHFFFAOYSA-N
octadecane-1-thiol
17905
96%

Chemical Identifiers

2885-00-9
286.56
QJAOYSPHSNGHNC-UHFFFAOYSA-N
17905
CCCCCCCCCCCCCCCCCCS
C18H38S
MFCD00004886
octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
octadecane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 220-744-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only