missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanesulfonic acid, sodium salt, 90%

CAS: 13893-34-0 | C18H37NaO3S | 356.54 g/mol

$386.10 - $386.10

Chemical Identifiers

CAS 13893-34-0
Molecular Formula C18H37NaO3S
Molecular Weight (g/mol) 356.54
MDL Number MFCD00051526
InChI Key KBAFDSIZQYCDPK-UHFFFAOYSA-M
Synonym sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
PubChem CID 23668003
IUPAC Name sodium;octadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384910100
SDS View Documents
Thermo Scientific Chemicals
384910100
10 g Glass bottle
Each for $386.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13893-34-0
356.54
KBAFDSIZQYCDPK-UHFFFAOYSA-M
23668003
CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
C18H37NaO3S
MFCD00051526
sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
sodium;octadecane-1-sulfonate

Specifications

13893-34-0
100.0
White
5% max. (1 G, 105°C, 1 Hrs, 760mm)
90%
C18H37NaO3S
MFCD00051526
sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
KBAFDSIZQYCDPK-UHFFFAOYSA-M
sodium;octadecane-1-sulfonate
23668003
90%
1-Octadecanesulfonic acid, sodium salt
88.0
163°C to 165°C
88% min. (C, H)
Authentic
Glass bottle
CH3(CH2)17SO3Na
10 g
Solubility in water: soluble
CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
356.54
356.54
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-660-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only