missing translation for 'onlineSavingsMsg'
Learn More

1-Octacosanol 93.0+%, TCI America™

Catalog No. O01991G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01991G 1 g
O0199100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01991G Supplier TCI America Supplier No. O01991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 557-61-9
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.77
MDL Number MFCD00044770
InChI Key CNNRPFQICPFDPO-UHFFFAOYSA-N
Synonym 1-octacosanol, octacosanol, montanyl alcohol, octacosyl alcohol, cluytyl alcohol, n-octacosanol, octacosanol-1, policosanol, octacosyl, polycosanol
PubChem CID 68406
ChEBI CHEBI:28243
IUPAC Name octacosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 410.77
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Octacosanol
EINECSNumber (2)-0217
TSCA Yes