missing translation for 'onlineSavingsMsg'
Learn More

1-Octacosanol, 90+%, Thermo Scientific™

Chemical Identifiers

CAS 557-61-9
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.77
MDL Number MFCD00044770
InChI Key CNNRPFQICPFDPO-UHFFFAOYSA-N
Synonym 1-octacosanol, octacosanol, montanyl alcohol, octacosyl alcohol, cluytyl alcohol, n-octacosanol, octacosanol-1, policosanol, octacosyl, polycosanol
PubChem CID 68406
ChEBI CHEBI:28243
IUPAC Name octacosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC258132500
View Documents Promotion Details
N/A 250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

557-61-9
410.77
CNNRPFQICPFDPO-UHFFFAOYSA-N
68406
octacosan-1-ol
C28H58O
MFCD00044770
1-octacosanol, octacosanol, montanyl alcohol, octacosyl alcohol, cluytyl alcohol, n-octacosanol, octacosanol-1, policosanol, octacosyl, polycosanol
CHEBI:28243
CCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

557-61-9
100.0
Authentic
C28H58O
MFCD00044770
1-octacosanol, octacosanol, montanyl alcohol, octacosyl alcohol, cluytyl alcohol, n-octacosanol, octacosanol-1, policosanol, octacosyl, polycosanol
CNNRPFQICPFDPO-UHFFFAOYSA-N
octacosan-1-ol
68406
410.77
1-Octacosanol
90.0
200.0°C to 250.0°C
90% min. (GC)
CH3(CH2)27OH
15, 6833
Solubility in water: insoluble. Other solubilities: soluble in hot hydrocarbons, hot dibutylether,, thf,,hot methylisobutylketone
CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
410.77
CHEBI:28243
90+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-181-2

SDS
Documents

Documents

RUO – Research Use Only