missing translation for 'onlineSavingsMsg'
Learn More

1-Nonyne 98.0+%, TCI America™

Supplier:  TCI America N03015ML

 View more versions of this product

Catalog No. N03015ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Nonyne
Colorless
C9H16
5 mL
1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci
CCCCCCCC#C
124.23
124.23
Liquid
9-3-3452
151°C
MFCD00009567
3295
OSSQSXOTMIGBCF-UHFFFAOYSA-N
non-1-yne
18937
≥98.0% (GC)

Chemical Identifiers

9-3-3452
124.23
OSSQSXOTMIGBCF-UHFFFAOYSA-N
18937
CCCCCCCC#C
C9H16
MFCD00009567
1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci
non-1-yne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions