missing translation for 'onlineSavingsMsg'
Learn More

1-Nonylamine, 97%

Catalog No. AAA1820914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1820914 25 g
AAA1820922 100 g
AAA1820936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1820914 Supplier Thermo Scientific Chemicals Supplier No. A1820914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-20-9 | C9H22ClN | 179.73 g/mol

1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 112-20-9
Molecular Formula C9H22ClN
Molecular Weight (g/mol) 179.73
MDL Number MFCD00008249
InChI Key IMGHCWMUWVHYCO-UHFFFAOYSA-N
Synonym nonylamine, n-nonylamine, 1-aminononane, 1-nonanamine, 1-nonylamine, unii-7l7h2ht4ok, 7l7h2ht4ok, nonyl-amine, n-nonyl amine, amine c9
PubChem CID 16215
IUPAC Name nonan-1-amine
SMILES [Cl-].CCCCCCCCC[NH3+]

Specifications

Melting Point -1°C
Density 0.784
Boiling Point 200°C to 202°C
Flash Point 62°C (143°F)
Odor Amine-like
Refractive Index 1.433
Quantity 25 g
UN Number UN2735
Beilstein 1733633
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 143.27
Percent Purity ≥98%
Chemical Name or Material 1-Nonylamine
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-945-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only