missing translation for 'onlineSavingsMsg'
Learn More

1-Nonene 95.0+%, TCI America™

Catalog No. N061325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N061325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N061325ML Supplier TCI America Supplier No. N061325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-11-8
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00009568
InChI Key JRZJOMJEPLMPRA-UHFFFAOYSA-N
PubChem CID 31285
ChEBI CHEBI:77443
IUPAC Name non-1-ene
SMILES CCCCCCCC=C

Specifications

Color Colorless
Boiling Point 147°C
Quantity 25 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nonene
EINECSNumber (2)-0025
TSCA Yes