missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol, 99%

Catalog No. p-7020525
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1251022 100 g
AAA1251036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1251022 Supplier Thermo Scientific Chemicals Supplier No. A1251022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143-08-8 | C9H20O | 144.258 g/mol

The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of nonanol is in the manufacture of artificial lemon oil. 1-Nonanol is mainly used in man-made rose essence, plasticizer, surfactant. 1-Nonanol was used as a reference material in the determination of airborne spore levels. 1-Nonanol inhibited the bacterial luciferase (BL) reaction in a dose-dependent manner.

Solubility
Soluble in water (1 g/L) at 20°C, ether, ethanol, acetone, chloroform, and mineral oil.

Notes
Store at room temperature. Ensure good ventilation.

Chemical Identifiers

CAS 143-08-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00002990
InChI Key ZWRUINPWMLAQRD-UHFFFAOYSA-N
Synonym 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
PubChem CID 8914
ChEBI CHEBI:35986
IUPAC Name nonan-1-ol
SMILES CCCCCCCCCO

Specifications

Melting Point -7°C
Density 0.827
Boiling Point 214°C to 216°C
Flash Point 75°C (167°F)
Odor Pleasant
Refractive Index 1.433
Linear Formula CH3(CH2)8OH
Quantity 100 g
UN Number UN3082
Beilstein 969213
Merck Index 14,6679
Solubility Information Soluble in water (1g/L) at 20°C,ether,ethanol,acetone,chloroform,and mineral oil.
Formula Weight 144.26
Percent Purity 99%
Chemical Name or Material 1-Nonanol
Show More Show Less
Hazard Category H227-H315-H319-H332-H335
Hazard Statement GHS H Statement
H332-H319-H227
Harmful if inhaled.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 205-583-7
RTECSNumber RB1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only