missing translation for 'onlineSavingsMsg'
Learn More

1-Nonadecanol 98.0+%, TCI America™

Catalog No. N02845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02845G 5 g
N028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02845G Supplier TCI America Supplier No. N02845G
Only null left

Chemical Identifiers

CAS 1454-84-8
Molecular Formula C19H40O
Molecular Weight (g/mol) 284.53
MDL Number MFCD00002824
InChI Key XGFDHKJUZCCPKQ-UHFFFAOYSA-N
Synonym 1-nonadecanol, nonadecanol, nonadecyl alcohol, unii-a465x576ko, n-nonadecanol-1, acmc-209cv8, 1-nonadecanol 10 ng/microl in methyl-tert. butyl ether, 1-nonadecanol, european pharmacopoeia ep reference standard
PubChem CID 80281
IUPAC Name nonadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 62°C
Color White
Quantity 5 g
Formula Weight 284.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nonadecanol
EINECSNumber (2)-0217
TSCA No