missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylmethylamine 98.0+%, TCI America™

Catalog No. p-7134100
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10425G 5 g
N104225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10425G Supplier TCI America Supplier No. N10425G

Chemical Identifiers

CAS 118-31-0
Molecular Formula C11H12N
Molecular Weight (g/mol) 158.22
MDL Number MFCD00004048
InChI Key NVSYANRBXPURRQ-UHFFFAOYSA-O
Synonym 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
PubChem CID 8355
IUPAC Name (naphthalen-1-yl)methanaminium
SMILES [NH3+]CC1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 293°C
Quantity 5 g
Formula Weight 157.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Naphthylmethylamine