missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylamine 99.0+%, TCI America™

Catalog No. p-7133596
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N005225G 25 g
N0052100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N005225G Supplier TCI America Supplier No. N005225G
Only null left

Chemical Identifiers

CAS 134-32-7
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00004016
InChI Key RUFPHBVGCFYCNW-UHFFFAOYSA-N
Synonym 1-naphthylamine, 1-aminonaphthalene, 1-naphthalenamine, alpha-naphthylamine, naphthalidine, naphthalen-1-ylamine, naphthalidam, 1-naphthalamine, naphthylamine, fast garnet base b
PubChem CID 8640
ChEBI CHEBI:50450
IUPAC Name naphthalen-1-amine
SMILES C1=CC=C2C(=C1)C=CC=C2N

Specifications

Melting Point 50°C
Color Red-White
Boiling Point 301°C
Quantity 25 g
UN Number 2077
Formula Weight 143.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthylamine
EINECSNumber (4)-0321
RTECSNumber QM1400000
TSCA Yes