missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthylamine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 50°C |
| Color | Red-White |
| Boiling Point | 301°C |
| Quantity | 25 g |
| UN Number | 2077 |
| Formula Weight | 143.19 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Naphthylamine |
Safety and Handling
| EINECSNumber | (4)-0321 |
| RTECSNumber | QM1400000 |
| TSCA | Yes |