missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylamine Hydrochloride 98.0+%, TCI America™

Catalog No. N005625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N005625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N005625G Supplier TCI America Supplier No. N005625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 552-46-5
Molecular Formula C10H10ClN
Molecular Weight (g/mol) 179.647
MDL Number MFCD00036370
InChI Key FOKKJVHTXPJHEN-UHFFFAOYSA-N
Synonym 1-naphthylamine hydrochloride, 1-naphthalenamine, hydrochloride, naphthalen-1-amine hydrochloride, 1-naphthylammonium chloride, 1-naphthalenamine hydrochloride, alpha-naphthylamine hydrochloride, 1-amino-naphthalene hydrochloride, 1-aminonaphthalene hcl, 1-naphthalenamine, hydrochloride 1:1, 1-aminonaphthalene hydrochloride
PubChem CID 11094
IUPAC Name naphthalen-1-amine;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2N.Cl

Specifications

Melting Point 275°C
Quantity 25 g
UN Number 2811
Formula Weight 179.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthylamine Hydrochloride
EINECSNumber (4)-0321
RTECSNumber QM3150000
TSCA Yes