missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetyl chloride, 95%

Catalog No. p-7033292
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5038603 1 g
AAH5038606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5038603 Supplier Thermo Scientific Chemicals Supplier No. H5038603
Only null left

CAS: 5121-00-6 | C12H9ClO | 204.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5121-00-6
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.65
MDL Number MFCD00175852
InChI Key DSVAZLXLRDXHKO-UHFFFAOYSA-N
Synonym 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride
PubChem CID 2795386
IUPAC Name 2-(naphthalen-1-yl)acetyl chloride
SMILES ClC(=O)CC1=CC=CC2=CC=CC=C12

Specifications

Density 1.23
Boiling Point 130°C (0.6 mmHg)
Odor Pungent
Refractive Index 1.621
Quantity 1 g
UN Number UN3265
Formula Weight 204.66
Percent Purity 95%
Chemical Name or Material 1-Naphthylacetyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only