missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetonitrile 98.0+%, TCI America™

Catalog No. N036625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N036625G 25 g
N0366500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N036625G Supplier TCI America Supplier No. N036625G

Chemical Identifiers

CAS 132-75-2
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.211
MDL Number MFCD00004041
InChI Key OQRMWUNUKVUHQO-UHFFFAOYSA-N
Synonym 1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile
PubChem CID 8596
IUPAC Name 2-naphthalen-1-ylacetonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2CC#N

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 167.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Naphthylacetonitrile
RTECSNumber AM1240000
TSCA Yes