missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylacetonitrile, 97%

Catalog No. AAL0669606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0669606 5 g
AAL0669614 25 g
AAL0669622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0669606 Supplier Thermo Scientific Chemicals Supplier No. L0669606

CAS: 132-75-2 | C12H9N | 167.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132-75-2
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.211
MDL Number MFCD00004041
InChI Key OQRMWUNUKVUHQO-UHFFFAOYSA-N
Synonym 1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile
PubChem CID 8596
IUPAC Name 2-naphthalen-1-ylacetonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2CC#N

Specifications

Melting Point 33°C to 35°C
Boiling Point 180°C to 182°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6192
Quantity 5 g
UN Number UN3439
Beilstein 1101012
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 1-Naphthylacetonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 205-078-1
RTECSNumber AM1240000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only