missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl Isocyanate 99.0+%, TCI America™

Catalog No. p-7131222
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01255ML 5 mL
I012525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01255ML Supplier TCI America Supplier No. I01255ML
Only null left

Chemical Identifiers

CAS 86-84-0
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.183
MDL Number MFCD00003881
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 269°C
Quantity 5 mL
UN Number 2206
Formula Weight 169.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Naphthyl Isocyanate
EINECSNumber (4)-0526
TSCA Yes
Recommended Storage Refrigerator