missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isocyanate, 98%

Catalog No. AAA1720014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1720014 25 g
AAA1720006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1720014 Supplier Thermo Scientific Chemicals Supplier No. A1720014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-84-0 | C11H7NO | 169.183 g/mol

1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isocyanate is used in the preparation of cationic aromatic urethane. It is useful for the study of cholangiolitic hepatotoxicity in laboratory animals. It is also employed as separate enantiomers of beta-blockers.

Solubility
Miscible with alcohol, chloroform and diethyl ether.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents and alcohols.

Chemical Identifiers

CAS 86-84-0
Molecular Formula C11H7NO
MDL Number MFCD00003881
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene

Specifications

Melting Point 4°C
Density 1.177
Boiling Point 269°C
Flash Point 135°C (275°F)
Odor Irritating
Refractive Index 1.6335
Quantity 25 g
UN Number UN2206
Beilstein 742779
Sensitivity Moisture sensitive
Merck Index 14,6410
Solubility Information Miscible with alcohol,chloroform and diethyl ether.
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O
Molecular Weight (g/mol) 169.183
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material 1-Naphthyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 201-703-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only