missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl Butyrate 98.0+%, TCI America™

Catalog No. B10965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10965G Supplier TCI America Supplier No. B10965G
Only null left

Chemical Identifiers

CAS 3121-70-8
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00003923
InChI Key XIVJNRXPRQKFRZ-UHFFFAOYSA-N
Synonym 1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
PubChem CID 76571
IUPAC Name naphthalen-1-yl butanoate
SMILES CCCC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

Boiling Point 182°C
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Naphthyl Butyrate